Sklarcut™ Stevens Tenotomy Scissors, OR Grade, Sklar
Supplier: Sklar
15-3370 15-3372
89025-572EA 438.41 CAD
89025-572 89025-574
Sklarcut™ Stevens Tenotomy Scissors, OR Grade, Sklar
Scissors
Sklarcut™ Stevens Tenotomy Scissors are used for delicate dissection and cutting, in ophthalmologic, neurological, and plastic surgery procedures
Tenotomy scissors have long handles and small sharp blades with sharp or blunt wedge-shaped tips. The long handles allow the surgeon to have an adequate grip and control while dissecting or cutting in small or narrow areas. Stevens Tenotomy Scissors are available straight or curved and with sharp or blunt tips to accommodate different procedures. Sharp tips allow precise dissection and cutting of delicate tissue, while blunt tipped blades are used for dissecting and cutting mucus membranes. Sklarcut™ scissors feature micro-serrations on one blade to reduce trauma when cutting both delicate and course tissue. These serrations assist in holding the tissue during cutting and minimizes tissue from slipping so the most accurate and clean cuts can be made. Sklarcut™ scissors are identified by a single gold coated finger ring on the handle of the serrated blade.