Pederson Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
90-3713 90-3711 90-3710 90-3712 90-3851 90-3849
10193-664EA 132.02 CAD
10193-664 10193-698 10193-804 10193-852 10193-948 10197-238
Pederson Vaginal Speculum, OR Grade, Sklar
Specula
Sklar® Pederson Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, pap smears, and examining the cervix. It is a double blade duckbill speculum inserted with the blades closed.
  • The blades are then opened by an external lever and screw
  • Reusable and latex-free
The blades of the Pederson speculum are more narrow than the blades of the Graves Speculum. The practitioner can best determine which type and size speculum is most appropriate for the patient.
Order Now
SPECIFICATIONS
Environmentally Preferable
Latex/Latex-free Latex-free