Sklar Blue™ Emmett Hook, OR Grade, Sklar
Supplier: Sklar
91-5250 91-5251 91-5252 91-5253 91-5254
10184-502PK 1830.74 CAD
10184-502 10184-504 10184-506 10184-508 10184-510
Sklar Blue™ Emmett Hook, OR Grade, Sklar
Dissection Needles
The Sklar Blue™ Emmett Hook, which may also be known as an Emmett Iris Hook, is most commonly used in electrosurgical OB/GYN procedures involving the uterus.
  • The hook comes in a variety of styles
  • Reusable
  • Latex-free
Sklar Blue™ instruments feature high quality stainless steel with an insulated coating that prevents radio frequency currents from being transmitted through the instrument and thus reducing the burning of non-targeted tissue. Sklar Blue™ instruments are fully autoclavable and may also be cleaned ultrasonically.