Anti-FOXP3 Rabbit Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1054C]

Supplier: R&D Systems
FPK8214P-025 FPK8214P-100
102884-122EA 395.38 CAD
102884-122 102884-124
Anti-FOXP3 Rabbit Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1054C]
Antibodies
Type: Primary
Antigen Symbol: FOXP3
Antigen Name: forkhead box P3
Antigen Synonyms: AIID, DIETER, IPEX, JM2, PIDX, XPID
Clonality: Monoclonal
Clone: 1054C
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR