Anti-TGFBR2 Goat Polyclonal Antibody (Fluorescein)

Supplier: R&D Systems
FAB532F-025 FAB532F-100
102882-900EA 296.92 CAD
102882-900 102882-902
Anti-TGFBR2 Goat Polyclonal Antibody (Fluorescein)
Antibodies
Type: Primary
Antigen: TGFBR2
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR