Anti-IL6ST Mouse Monoclonal Antibody (Fluorescein) [clone: 28126]

Supplier: R&D Systems
FAB228F-025 FAB228F-100
102881-978EA 315.38 CAD
102881-978 102881-980
Anti-IL6ST Mouse Monoclonal Antibody (Fluorescein) [clone: 28126]
Antibodies
Type: Primary
Antigen: IL6ST
Clonality: Monoclonal
Clone: 28126
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR