Anti-IL12RB2 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 305719]

Supplier: R&D Systems
FAB1959C-025 FAB1959C-100
102881-592EA 416.92 CAD
102881-592 102881-594
Anti-IL12RB2 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 305719]
Antibodies
Type: Primary
Antigen: IL12RB2
Clonality: Monoclonal
Clone: 305719
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR