Anti-IL17RB Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 752101]

Supplier: R&D Systems
FAB10402A-025 FAB10402A-100
102880-790EA 416.92 CAD
102880-790 102880-792
Anti-IL17RB Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 752101]
Antibodies
Type: Primary
Antigen: IL17RB
Clonality: Monoclonal
Clone: 752101
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR