Sklar Blue™ Emmett Hook, Quick Release , OR Grade, Sklar
Supplier: Sklar
91-5241 91-5242 91-5245 91-5248
10184-492EA 640.48 CAD
10184-492 10184-494 10184-496 10184-500
Sklar Blue™ Emmett Hook, Quick Release , OR Grade, Sklar
Retractors
The Sklar Blue™ Emmett Hook, which may also be known as an Emmett Iris Hook, is most commonly used in electrosurgical OB/GYN procedures involving the uterus.
  • Fully autoclavable and may also be cleaned ultrasonically
  • Available in multiple diameters
The hook is at the end of a long, narrow handle. Sklar Blue™ instruments feature high quality stainless steel with an insulated coating that prevents radio frequency currents from being transmitted through the instrument and thus reducing the burning of non-targeted tissue.