Sklar Blue™ Freeway-Graves Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
91-5160 91-5162
10184-468EA 933.45 CAD
10184-468 10184-470
Sklar Blue™ Freeway-Graves Vaginal Speculum, OR Grade, Sklar
Specula
The Sklar Blue™ Freeway-Graves Vaginal Speculum is used for electrosurgical gynecological procedures.
  • Fully autoclavable
  • Can be cleaned ultrasonically
It is available with or without a permanent smoke evacuation tube for use during cauterization. The Freeway-Graves Vaginal Speculum features an open side which allows improved visibility and more room for supplemental instrument access. It is available with 45 or 90 degree angled blades. It feature high quality stainless steel with an insulated coating.
This safe coating prevents radio frequency currents from being transmitted through the instrument which reduces the burning of non-targeted tissue. Sklar Blue™ electrosurgical coatings are guaranteed for one year against manufacturing defects.
Certifications: All Sklar Blue™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79.