Sklar Blue™ Cusco Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
91-5150 91-5152 91-5154
10184-462EA 752.57 CAD
10184-462 10184-464 10184-466
Sklar Blue™ Cusco Vaginal Speculum, OR Grade, Sklar
Specula
The Cusco vaginal speculum with lateral screw is a self-retaining speculum used for examination of, or procedures within, the vaginal canal.
  • The blades of the Cusco speculum are opened, closed, and adjusted with the attached screw
  • Electrosurgical instrument feature high quality stainless steel with an insulated coating
This safe coating prevents radio frequency currents from being transmitted through the instrument which reduces the burning of non-targeted tissue.