Oblong Plastic Bottles, Narrow, Mouth, HDPE, Natural, C&G Containers
Supplier: C&G Containers
LPO004200010 LPO004900000 LPO004900001 LPO008200012 LPO008900000 LPO016900000 LPO032200013
71001-012CS 1211.5 CAD
71001-012 71001-016 71001-018 71001-022 71001-024 71001-028 71001-032 71001-038
Oblong Plastic Bottles, Narrow, Mouth, HDPE, Natural, C&G Containers
Bottles Plastic Bottles
Analysis for certified analytes and corresponding reporting limits.
Pre-cleaned and Certified products contain the Certificate of Analysis and Cleaning Protocol Form in the product case.
A lot certificate of analysis is generated with the analytical results obtained, including the concentration reporting limits and units of measurement, for Pre-cleaned and Certified products. This certificate of analysis is included in each product case.
Wash protocols for these bottles include; A1 (Nitric acid wash), B1 (Deionized water wash), C1 (Hydrochloric acid wash), and L3 (no wash).