CENCO® Spectrometer Prisms
Supplier: Wards
PH0583B PH0583C PH0583A PH0583D PH0583E
470219-500EA 81.3 CAD
470219-500 470219-502 470219-498 470219-504 470219-506
CENCO® Spectrometer Prisms
Educational Materials Physics Educational Materials Optics Learning Activities
Spectrometer prisms offer a different index of refraction for every need.
  • Different Indices of Refractions for Every Need
  • Glass Material
  • Designed to Yield Accurate Results
Although any prism can be used in a spectrometer, these are specially designed to yield accurate results for spectroscopy experiments.
Choose your prism type and the approximate index of refraction that would work best for your experiment.