Manometer Demonstration
Supplier: Wards
PH0203G
470219-420EA 145.5 CAD
470219-420
Manometer Demonstration
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
Demonstrate the effect of pressure to water depth.
  • Complete package to highlight the variation of pressure in a liquid with depth
It consists of a pressure capsule of 35mm diameter placed in a bent steel tube. The pressure exerted by the water on the membrane of the capsule is measured by a pressure gauge mounted on a graduated support, connected to the probe by means of a transparent flexible tube. A device used to turn the capsule without leaving the water. The manometer tube is slideable on the support to facilitate setting of zero. Supplied with beaker.
Order Now
SPECIFICATIONS
Environmentally Preferable