BD Vacutainer® Blood Collection Needles, BD Diagnostics
Supplier: BD
360212 360210 360214 360213
CABD360212CS 401.31 CAD
CABD360212 CABD360210 CABD360214 CABD360213L
BD Vacutainer® Blood Collection Needles, BD Diagnostics
Syringe Needles
BD Vacutainer® needles can be used for multiple tube samples
They have an advanced low-angle bevel design and are coated with silicone, a low friction lubricant, ensuring very gentle and smooth vein entry. Individual laser quality control maintains the highest quality levels. Available with a 20, 21 or 22 gauge needle.
Order Now
SPECIFICATIONS
Environmentally Preferable