Versette™ Automated Liquid Handler, Thermo Scientific
Supplier: Thermo Fisher Scientific
650-01-BS 650-02-NTC 650-03-SPS 650-06-9630 650-06-96300 650-06-384100
CA97054-038EA 93475.52 CAD
CA97054-038 CA97054-042 CA97054-048 CA97054-060 CA97054-062 CA97054-066
Versette™ Automated Liquid Handler, Thermo Scientific
Automated Liquid Handling Systems
Versette™ is a compact automated liquid handler featuring user-friendly programming combined with reliable performance, which makes it an ideal choice for the liquid handling needs of most laboratories.
  • Equipped with a safety shield to ensure maximal user safety
  • Simple pipetting procedures can be created using visual on-board graphical display
  • Fits on standard bench and biological hoods
The Versette system is compatible with disposable automation research tips (D.A.R.T.S™), which feature a unique surface seal design to ensure an even seal across allMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable