Flame Resistant Long Sleeve Henley Shirt with Max Comfort™ Material, Navy Blue, MCR Safety
Supplier: MCR Safety
H1NL H1NM H1NS H1NX2 H1NX3 H1NX4 H1NX5 H1NXL
10813-470EA 208.56 CAD
10813-470 10813-472 10813-474 10813-476 10813-478 10813-480 10813-482 10813-484
Flame Resistant Long Sleeve Henley Shirt with Max Comfort™ Material, Navy Blue, MCR Safety
Workwear
Whether working in the rugged oil and gas industry or spending the day working near electrical hazards, stay safe and comfortable in MCR Safety's Flame Resistant (FR) Gear made with Max Comfort™ fabrics
  • 100% FR Cotton
  • Left chest pocket
  • Moisture wicking and breathable
  • Rib-knit collar and cuff
  • NFPA 2112, NFPA 70E, UL Certified
The H1 Henley Shirt features a 100% FR cotton material for maximum comfort. The Max Comfort™ material meets the NFPA 2112-2012 Standard for Flame Resistant Garments and NFPA 70E Standard for Electrical Safety in the Workplace. All of MCR Safety's FR Garments are UL Certified and have a lifetime FR guarantee.
Certifications: NFPA 2112, NFPA 70E, UL Certified.