DuPont™ Tychem® 4000 Coveralls with Attached Socks, Bound Seams
Supplier: DuPont
SL121BWH2X001200 SL121BWH3X001200 SL121BWH4X001200 SL121BWH5X001200 SL121BWH6X001200 SL121BWHLG001200 SL121BWHMD001200 SL121BWHXL001200
10016-432CS 957.9 CAD
10016-432 10016-434 10016-436 10016-438 10016-440 10016-442 10016-444 10016-446
DuPont™ Tychem® 4000 Coveralls with Attached Socks, Bound Seams
Disposable Garments
DuPont™ Tychem® 4000 delivers effective protection against a range of chemical environments.
  • Laydown collar
  • Elastic wrists
  • Attached socks
  • Storm flap with adhesive closure
  • Bound seams
Tychem® 4000, utilizes Saranex™ 23-P film laminated to Tyvek® fabric to provide a rugged and durable fabric that resists abrasion. Tychem® 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications include chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments.More Product Information