Avantor® Seradigm, Ultra Low IgG Fetal Bovine Serum (FBS)

Supplier: VWR
4100-500 4100-500H 4100-500G 4100-500GH
10018-826EA 1557.61 CAD
10018-826 10018-830 10836-696 10836-698
Avantor® Seradigm, Ultra Low IgG Fetal Bovine Serum (FBS)
Sera (Serum)
Ultra Low IgG Fetal Bovine Serum (FBS) is purified of immunoglobulin (IgG) using a chromatographic process that delivers significantly reduced levels of IgG (≤5μg/mL).

  • 100% US Origin
  • IgG level ≤5μg/mL
  • Ideal for cell culture and protein purification applications where naturally occurring levels of IgG are too high
  • ISIA Certified Supply Chain for Traceability

Ultra Low IgG Fetal Bovine Serum (FBS) is ideal for cell culture and protein purification applications where naturally occurring levels of IgG are too high.

Avantor Seradigm is a supplier ofMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Endotoxin ≤ 5 EU/mL
Hemoglobin ≤ 25 mg/dL
Storage temperature Frozen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR