DuPont™ ProShield® 10 Coveralls with Laydown Collar and Elastic Wrists
Supplier: DuPont
PB125SBU2X002500 PB125SBU3X002500 PB125SBU4X002500 PB125SBU5X002500 PB125SBU6X002500 PB125SBU7X002500 PB125SBULG002500 PB125SBUMD002500 PB125SBUSM002500 PB125SBUXL002500 PB125SWH2X002500 PB125SWH3X002500 PB125SWH4X002500 PB125SWH5X002500 PB125SWH6X002500 PB125SWH7X002500 PB125SWHLG002500 PB125SWHMD002500 PB125SWHSM002500
10016-222CS 174.58 CAD
10016-222 10016-224 10016-226 10016-228 10016-230 10016-232 10016-234 10016-236 10016-238 10016-240 10016-242 10016-244 10016-246 10016-248 10016-250 10016-252 10016-254 10016-256 10016-258 10016-260
DuPont™ ProShield® 10 Coveralls with Laydown Collar and Elastic Wrists
Disposable Garments
DuPont™ ProShield® 10, spunbond meltblown spunbond (SMS) garments, are made to the high quality standards you have come to expect from DuPont.
  • Laydown collar
  • Elastic wrists and ankles
  • Elastic waist
  • Serged seams
ProShield® 10 garments have been optimized for comfort, softness and breathability while enabling you to cut costs for disposable garments. DuPont™ ProShield® 10 disposable garments are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications. Applications include general maintenance, janitorial/cleaning and other dirty work. Coveralls feature a laydown collar and elastic openings at wrists and ankles for a tighter fit.
Certifications: Thickness meets ASTM D1117.