DuPont™ ProShield® 10 Coveralls with Attached Hood and Boots
Supplier: DuPont
PB122SBU2X002500 PB122SBU3X002500 PB122SBU4X002500 PB122SBU5X002500 PB122SBU6X002500 PB122SBU7X002500 PB122SBULG002500 PB122SBUMD002500 PB122SBUSM002500 PB122SBUXL002500 PB122SWH2X002500 PB122SWH3X002500 PB122SWH4X002500 PB122SWH5X002500 PB122SWH6X002500 PB122SWH7X002500 PB122SWHLG002500 PB122SWHMD002500 PB122SWHSM002500 PB122SWHXL002500
10016-182CS 377.55 CAD
10016-182 10016-184 10016-186 10016-188 10016-190 10016-192 10016-194 10016-196 10016-198 10016-200 10016-202 10016-204 10016-206 10016-208 10016-210 10016-212 10016-214 10016-216 10016-218 10016-220
DuPont™ ProShield® 10 Coveralls with Attached Hood and Boots
Disposable Garments
DuPont™ ProShield® 10, spunbond meltblown spunbond (SMS) garments, are made to the high quality standards you have come to expect from DuPont.
  • Standard fit hood
  • Elastic wrists
  • Attached skid-resistant boots
  • Elastic waist
  • Serged seams
ProShield® 10 garments have been optimized for comfort, softness and breathability while enabling you to cut costs for disposable garments. DuPont™ ProShield® 10 disposable garments are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications. Applications include general maintenance, janitorial/cleaning and other dirty work. Attached hood with elastic around face opening is a standard fit hood. Elastic openings at wrists allows for a tighter fit. Attached boots feature skid-resistant soles.
Certifications: Thickness meets ASTM D1117.