DuPont™ ProShield® 10 Coveralls with Laydown Collar and Open Wrists
Supplier: DuPont
PB120SBU5X002500 PB120SBU6X002500 PB120SBU7X002500 PB120SBULG002500 PB120SBUMD002500 PB120SBUSM002500 PB120SBUXL002500 PB120SWH2X002500 PB120SWH3X002500 PB120SWH4X002500 PB120SWH5X002500 PB120SWH6X002500 PB120SWHLG002500 PB120SWHMD002500 PB120SWHSM002500 PB120SWHXL002500 PB120SBU2X002500 PB120SBU3X002500 PB120SBU4X002500
10016-148CS 163.3 CAD
10016-148 10016-150 10016-152 10016-154 10016-156 10016-158 10016-160 10016-162 10016-164 10016-166 10016-168 10016-170 10016-172 10016-174 10016-176 10016-178 10016-180 10016-742 10016-744 10016-746
DuPont™ ProShield® 10 Coveralls with Laydown Collar and Open Wrists
Disposable Garments
DuPont™ ProShield® 10, spunbond meltblown spunbond (SMS) garments, are made to the high quality standards you have come to expect from DuPont.
  • Laydown collar
  • Open wrists and ankles
  • Elastic waist
  • Serged seams
ProShield® 10 garments have been optimized for comfort, softness and breathability while enabling you to cut costs for disposable garments. DuPont™ ProShield® 10 disposable garments are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications. Applications include general maintenance, janitorial/cleaning and other dirty work. Coveralls feature a laydown collar and open wrists and ankles.
Certifications: Thickness meets ASTM D1117.