DuPont™ ProShield® 60 Coveralls with Respirator Hood
Supplier: DuPont
NG127SWH2X0025NP NG127SWH3X0025NP NG127SWH4X0025NP NG127SWH5X0025NP NG127SWH6X0025NP NG127SWHLG0025NP NG127SWHMD0025NP NG127SWHSM0025NP NG127SWHXL0025NP
10016-108CS 359.37 CAD
10016-108 10016-110 10016-112 10016-114 10016-116 10016-118 10016-120 10016-122 10016-124
DuPont™ ProShield® 60 Coveralls with Respirator Hood
Disposable Garments
ProShield® 60 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
  • Respirator fit hood
  • Elastic wrists and ankles
  • Storm flap
  • Serged seams
ProShield® 60 garments are made with a microporous film that is laminated to a spundbonded polypropylene substrate. Applications include automotive refinishing, waste cleanup and remediation. Attached respirator-fit hood with elastic around face opening was designed to cover neck and chin and fit around respirator face mask. Longer zipper extends to chin for complete coverage of neck area. Storm flap of garment material covers zipper closure. Elastic openings at wrists and ankles allows for a tighter fit.
Certifications: Thickness meets ASTM D751.