QualaSheer® VHC12 QualaSheer XC Cleanroom Vinyl 12" Gloves, QRP
Supplier: PIP
VHC12S VHC12M VHC12XL
89040-956CS 723.7 CAD
89040-956 89040-958 89040-962
QualaSheer® VHC12 QualaSheer XC Cleanroom Vinyl 12" Gloves, QRP
Gloves
The QRP VHC12 ESD-Safe Clean Room Class 100 Vinyl ISO 5 Gloves are 100% PVC co-polymer (no latex) for ISO 5 (Class 100) applications.
  • Naturally anti-static; 10¹⁰ ohms
  • Microtextured
Certifications: Meets or exceeds all applicable requirements of IEST-RP-CC005.2.
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable
Latex/Latex-free Latex
Powdered/Powder-free Powder-free
Cleanroom class 100/ISO 5