SFX250 Sonifier® Cell Disruptor, with 1/2" Horn, Branson Ultrasonics
Supplier: Branson
Sonifier®
101-063-965R
10820-440EA 10633.67 CAD
10820-440
SFX250 Sonifier® Cell Disruptor, with 1/2" Horn, Branson Ultrasonics
Homogenizers Ultrasonic Cell Disruptors
The Sonifier® SFX250 is designed to bring the industry's most advanced sample-processing capabilities into any laboratory
True temperature control allows the user to specify a temperature range for the sample. The Sonifier® will automatically adjust the ultrasonic pulse length to prevent the sample from overheating (requires optional temperature probe).
Advanced control modes include time and energy mode, energy mode measured in joules, either continuously or in individual pulses. Process monitoring on a scrollable, digital screen provides updates of key variables (including power level, energy usage, sample temperature, and experiment progress). For assured repeatability and precision on a greater scale, users can store up to 20 sample-processing programs. Program parameters include continuous or pulsed ultrasonics; time, energy, or temperature control modes; pulse on-time/energy, off-time and total on-time/energy; or amplitude as a percentage. Microtip limit is 70%. Other accessories are also available separately.
Applications: Sample preparation, cell disruption, nanoparticle disperson, mixing, homogenization, DNA, cell lysis, protein extration, and shear/chip assays.
Able to accommodate sample volumes ranging up to 500mL, the Sonifier® SFX250 delivers up to 250 watts of power at 20kHz and comes with a 1/2" disruptor horn. The Sonifier® SFX250 offers the industry's simplest user interface and the most precise control over all the critical elements of any experiment.
Certifications: CSA, UL, ISO9001, ISO14001.
Accessories information: The Sound proof enclosure reduces noise level up to 20db when liquids are treated ultrasonically. Especially helpful when using a cell disrupter for extended periods.
Ordering information: The Sonifier® Cell Disruptor is supplied with a manufacturer's three-year warranty. The unit also includes a 20kHz converter, 1/2" tapped disruptor horn, power cord, manual, and wrench set. Stand and clamp are not included and sold separately.