Scout® Portable Topload Balance with Square Pan and Touchscreen Display, OHAUS®
Supplier: Ohaus
30253016 30253010 30253011 30253012 30253013 30253014 30253015
10805-276EA 1169.2 CAD
10805-276 10805-284 10805-286 10805-288 10805-290 10805-292 10805-294
Scout® Portable Topload Balance with Square Pan and Touchscreen Display, OHAUS®
Balances and Scales
Ideal for laboratory and industrial applications, the OHAUS® Scout® features an easy to navigate color touchscreen display and a slim design which allows stackable storage. Geared for superior performance with fast stabilization time and high resolution weighing results, these portable balances set a new standard for a broad range of weighing applications. Features also include superior overload protection and expanded connectivity options.
  • Informative Color Touchscreen Display
  • Superior Overload Protection and Stackable Storage
Additional standard features include; transportation lock, stainless steel pan, menu and calibration lockout switch, slip-resistant and adjustable feet, leveling bubble, mechanical and software overload/underload protection, stability indicator, auto tare, low battery indicator, auto shut-off, user selectable printing options, and user selectable communication settings.
Applications include Weighing, Parts Counting, Percent Weighing, Check Weighing, Animal/Dynamic Weighing, Totalization, Density Determination, Display Hold, and Mole Weighing.
Note: Model STX421 has a round pan, all other models have a square pan.
Certifications: IEC/EN 61010-1; CAN/CSA C22.2 No. 61010-1; UL Std. No. 61010-1, IEC/EN 61326-1 Class B; FCC Part 15 Class B; Industry Canada ICES-003 Class B.
Ordering information: Includes AC adapter (4 AA batteries not included).