Scout® Portable Topload Balance with Draftshield and Touchscreen Display, OHAUS®
Supplier: Ohaus
30253005 30253006
10805-358EA 1813.81 CAD
10805-358 10805-360
Scout® Portable Topload Balance with Draftshield and Touchscreen Display, OHAUS®
Balances and Scales
Ideal for laboratory and industrial applications, the OHAUS® Scout® features an easy to navigate color touchscreen display and a slim design which allows stackable storage. Geared for superior performance with fast stabilization time and high resolution weighing results, these portable balances set a new standard for a broad range of weighing applications. Features also include superior overload protection and expanded connectivity options.
  • Informative Color Touchscreen Display
  • Superior Overload Protection and Stackable Storage
Additional standard features include; transportation lock, stainless steel pan, menu and calibration lockout switch, slip-resistant and adjustable feet, leveling bubble, mechanical and software overload/underload protection, stability indicator, auto tare, low battery indicator, auto shut-off, user selectable printing options, and user selectable communication settings.
Applications include Weighing, Parts Counting, Percent Weighing, Check Weighing, Animal/Dynamic Weighing, Totalization, Density Determination, Display Hold, and Mole Weighing.
Ordering information: Includes AC adapter (4 AA batteries not included).