Scout® Portable Topload Balance with Draftshield, OHAUS®
Supplier: Ohaus
30253018 30253017
10805-278EA 1783.09 CAD
10805-278 10812-426
Scout® Portable Topload Balance with Draftshield, OHAUS®
Balances and Scales
Ideal for laboratory and industrial applications, the OHAUS® Scout® comes in a slim, stackable design with large backlit LCD. Features include superior overload protection, multiple weighing units, and application modes. Geared for high performance in any facility with fast stabilization time and high resolution weighing results, these portable balances set a new standard in laboratory and industrial weighing.
  • Bright Backlit LCD and 4-Button Control
  • Superior Overload Protection and Impact-Resistant Pan Support
Additional standard features include; transportation lock, stainless steel pan, menu and calibration lockout switch, slip-resistant and adjustable feet, leveling bubble, mechanical and software overload/underload protection, stability indicator, auto tare, low battery indicator, auto shut-off, user selectable printing options, and user selectable communication settings.
Applications include Weighing, Parts Counting, Percent Weighing, Check Weighing, Animal/Dynamic Weighing, Totalization, Density Determination, Display Hold, and Mole Weighing.
Certifications: IEC/EN 61010-1; CAN/CSA C22.2 No. 61010-1; UL Std. No. 61010-1, IEC/EN 61326-1 Class B; FCC Part 15 Class B; Industry Canada ICES-003 Class B.
Ordering information: Includes AC adapter (4 AA batteries not included).