Ranger® 4000 Compact Industrial Counting Scales, OHAUS®
Supplier: Ohaus
30236942 30236779 30236940 30236941
10820-640EA 2326.41 CAD
10820-640 10820-784 10820-786 10820-788
Ranger® 4000 Compact Industrial Counting Scales, OHAUS®
Balances and Scales
Ranger® 4000 Compact Industrial Counting Scales offer the best value for durable industrial weighing and counting applications. The largest, white LED display on any scale in its class, combined with checkweighing LEDs and audible indicators, make the Ranger® 4000 highly functional in loud and dim industrial settings. The high resolution Ranger® Count 4000 has a 1:30,000 display resolution and 1:1,500,000 internal counting resolution. It also features advanced auto-optimization software which recalculates the average piece weight as the overall weight increases, ensuring accuracy and minimizing counting errors.
  • 100-hours continuous use (12-hour recharge time)
  • Internal rechargeable lead acid battery
All models feature a stainless steel platform, integral weigh-below hook, sealed front panel, menu lock switch, up-front level indicator, adjustable leveling feet, selectable environmental and auto-print settings, stability indicator, overload and underload indicators, low battery indicator, auto shut-off, and auto tare.
Additional standard features include; die cast aluminum housing, a variety of weighing units (kg, g, lb, oz, lb:oz), a variety of application modes (Weighing, Percent Weighing, Parts Counting, Checkweighing, Checkcounting, Checkpercent, Accumulation, Dynamic Weighing/Display Hold), and a 40mm (1.5") white LED display with 6-digit, 7-segment capabilities.
Certifications: OIML R76 (R76/2006-NL1-15.21); NTEP (CC 13-022); Measurement Canada (AM-5907C), IEC/EN 60950-1; CAN/CSA C22.2 60950-1; UL 60950-1, IEC/EN 61326-1 Class B, Basic environments; FCC Part 15 Class B; Industry Canada ICES-003 Class B, WEEE/ROHS.