Ranger® 4000 Compact Industrial Scales, OHAUS®
Supplier: Ohaus
30236775 30236776 30236777 30236778
10820-776EA 2208.52 CAD
10820-776 10820-778 10820-780 10820-782
Ranger® 4000 Compact Industrial Scales, OHAUS®
Balances and Scales
Ranger® 4000 Compact Industrial Scales offer the best value for durable industrial weighing applications. These units feature the largest, white LED display on any scale in its class. Combined with checkweighing LEDs and audible indicators, this makes the Ranger® 4000 highly functional in loud and dim industrial settings.
  • 100-hours continuous use (12-hour recharge time)
  • Internal rechargeable lead acid battery
All models feature a stainless steel platform, integral weigh-below hook, sealed front panel, menu lock switch, up-front level indicator, adjustable leveling feet, selectable environmental and auto-print settings, stability indicator, overload and underload indicators, low battery indicator, auto shut-off, and auto tare.
Additional standard features include; die cast aluminum housing, a variety of weighing units (kg, g, lb, oz, lb:oz), a variety of application modes (Weighing, Percent Weighing, Parts Counting, Checkweighing, Checkcounting, Checkpercent, Accumulation, Dynamic Weighing/Display Hold), and a 40mm (1.5") white LED display with 6-digit, 7-segment capabilities.
Certifications: OIML R76 (R76/2006-NL1-15.21); NTEP (CC 13-022); Measurement Canada (AM-5907C), IEC/EN 60950-1; CAN/CSA C22.2 60950-1; UL 60950-1, IEC/EN 61326-1 Class B, Basic environments; FCC Part 15 Class B; Industry Canada ICES-003 Class B, WEEE/ROHS.