Petite Digital Stirrer Kit, Caframo
Supplier: Caframo
BDC250 BDC250MU1 BDC250U1
CA97001-138EA 704.28 CAD
CA97001-138 CA10860-292 CA10860-294
Petite Digital Stirrer Kit, Caframo
Stirrers Overhead Stirrers
The Petite Digital Stirrer Kit is supplied complete and ready to use.
Offering a significant amount of power in a small package, the Petite Digital Stirrer can be used in a variety of applications, such as R&D, clinical research, and educational labs. The compact footprint allows the stirrer to fit into small spaces and transport easily. The unit is designed to mix viscosities equivalent to water.
Certifications: UL listed. CSA approved. FCC, CE, IEC1010, RoHS, and WEEE compliant.
Ordering information: This stirrer is supplied with a three-year manufacturer's warranty.