Impeller Shaft, 316L Stainless Steel, Caframo
Supplier: Caframo
A162 A162 A712 A713 A722 A723 A733 A742 A743 A753 A712 A713 A722 A723 A733 A742 A743 A753
CA27800-362EA 111.6 CAD
CA27800-362 52428-752 52428-794 52428-796 52428-798 52428-800 52428-802 52428-804 52428-806 52428-808 CA52428-794 CA52428-796 CA52428-798 CA52428-800 CA52428-802 CA52428-804 CA52428-806 CA52428-808
Impeller Shaft, 316L Stainless Steel, Caframo
Caframo Impeller Shafts are composed of 316L electropolished stainless steel
Caframo 3/8" diameter shafts are turned down on one end to 1/2 x 5/16" diameter to accommodate 5/16" impeller bores. The 3/8" diameter shafts can be used with any impeller with a bore size of 5/16" or 3/8".
Offering a range of lengths in 5/16" or 3/8" diameters.