Pitched Blade Propellers, with Set Screws, Caframo
Supplier: Caframo
A511 A521 A531 A533 A541 A511 A521 A531 A533 A541
52428-780EA 70.27 CAD
52428-780 52428-782 52428-784 52428-786 52428-788 CA52428-780 CA52428-782 CA52428-784 CA52428-786 CA52428-788
Pitched Blade Propellers, with Set Screws, Caframo
Stirrers Stirring Tools
Caframo Pitched Blade Propellers are composed of 316L electropolished stainless steel
They offer low shear, high top-to-bottom pumping action with high gas (air) injection. Axial flow. Set screws included.
Ordering information: Shaft sold separately. See the Impeller Shaft, 316L Stainless Steel, Caframo series.