Dispersion Blade Impellers
Supplier: Caframo
A164 A164 A431 A441
52428-778EA 366.18 CAD
52428-778 CA52428-778 CA77860-240 CA77860-242
Dispersion Blade Impellers
Stirrers Stirring Tools
Caframo Dispersion Blade Impellers are electropolished 316 stainless steel dispersion blades ideal for high shear mixing at medium to high speeds, include blade only.
  • Designed with a uniform bore (8 mm) compatible with all Caframo shafts
  • Available in multiple sizes
  • Quick changes to test and optimize different batch sizes
  • Chemically inert and corrosion resistant
  • Broken particles are efficiently dispersed
When mixing, particles hit the blades and are broken apart and dispersed by the laminar flow set up by the blade.
52428-778 features a 1.9" diameter with 5/16" bore, and includes 4 set screws and Allen key. 
Ordering information: Shaft available separately.
Delivery information:
Supplied with set screws.