Collapsible Blade Impeller, 3.5", Caframo
Supplier: Caframo
A141 A141
52428-774EA 160.8 CAD
52428-774 CA58960-145
Collapsible Blade Impeller, 3.5", Caframo
Stirrers Stirring Tools
The Caframo Collapsible Blade Impeller features a 3.5" diameter, and is pinned to a 5/16" by 16" shaft
The impeller will vortex 5 gallons of water at 100rpm. Radial flow.
Composed of 316L electropolished stainless steel, it offers high shear for mixing in carboys or narrow neck flasks, and will fit into an opening greater than 0.4".