Plastic Packers, Wide Mouth, HDPE, Natural, C&G Containers
Supplier: C&G Containers
LPN128900002 LPW002200010 LPW002200011 LPW002900000 LPW004200011 LPW004900000 LPW008200012 LPW008220012 LPW008230012 LPW008900000 LPW008900002 LPW016200010 LPW016200012 LPW016220012 LPW016230012 LPW016900002 LPW032200013 LPW032230013 LPW032900003 LPW064200010 LPW064900004 LPW066200010 LPW128200010 LPW128900000
71001-010CS 46.33 CAD
71001-010 71001-106 71001-108 71001-110 71001-114 71001-116 71001-118 71001-122 71001-124 71001-128 71001-130 71001-132 71001-134 71001-136 71001-138 71001-140 71001-142 71001-144 71001-146 71001-148 71001-152 71001-158 71001-160 71001-162 71001-164 71001-166 71001-170 71001-174
Plastic Packers, Wide Mouth, HDPE, Natural, C&G Containers
Bottles Water Sampling Bottles
These plastic bottles are used for the collection of aqueous media for testing metals and a variety of inorganic parameters, including alkalinity, anions, acidity, cyanide, sulfate, and hardness.
Pre-cleaned and Certified products contain the Certificate of Analysis and Cleaning Protocol Form in the product case.
A lot certificate of analysis is generated with the analytical results obtained, including the concentration reporting limits and units of measurement, for Pre-cleaned and Certified products. This certificate of analysis is included in each product case.
Wash protocols for these bottles include; A1 (Nitric acid wash), B1 (Deionized water wash), C1 (Hydrochloric acid wash), and L3 (no wash). Some items are chemically preserved.
Not all analytical parameters are included on the Certificate of Analysis. Please refer to the Certificate of Analysis for certified analytes and corresponding reporting limits. Secondary uses for these containers include containment and archiving. Please refer to the applicable Certificate of Analysis for certified analytes and corresponding reporting limits.
Ordering information: Container measurements and capacity 'contain' volumes may vary.