Tungsten Carbide Allis Forceps, OR Grade, Sklar
Supplier: Sklar
97-1555 97-1560
10184-774EA 808.03 CAD
10184-774 10184-776
Tungsten Carbide Allis Forceps, OR Grade, Sklar
Forceps
Tungsten Carbide Atraumatic Allis Tissue Forceps are finger ring, ratcheted forceps used for grasping organs and slippery or dense tissue
Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.
TC instruments are identified by gold handles.
They are used with tonsil, vaginal, breast, and thyroid tissue. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments.