Sklar Blue™ Potts-Smith Dressing Forceps, OR Grade, Sklar
Supplier: Sklar
91-5411 91-5412 91-5414
10184-532EA 569.13 CAD
10184-532 10184-534 10184-536
Sklar Blue™ Potts-Smith Dressing Forceps, OR Grade, Sklar
Forceps
Sklar Blue™ Potts-Smith Dressing Forceps are most commonly used for application and changing of dressings in electrosurgical procedures
Serrated tips provide a solid grip on both materials and tissue. Sklar Blue™ electrosurgical instruments feature high quality stainless steel with an insulated coating. This safe coating prevents radio frequency currents from being transmitted through the instrument which reduces the burning of non-targeted tissue.
Sklar Blue™ instruments are fully autoclavable and may also be cleaned ultrasonically.
All Sklar Blue™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. Sklar Blue™ electrosurgical coatings are guaranteed for one year against manufacturing defects.
They may also be used for dissection of fine or single-layer tissue.