Econo™ Sterile Kelly Hemostatic Forceps, Floor Grade, Sklar
Supplier: Sklar
96-2560M 96-2562M 96-2561M 96-2563M
10022-492CS 1511.66 CAD
10022-492 10022-494 75995-400 75995-402
Econo™ Sterile Kelly Hemostatic Forceps, Floor Grade, Sklar
Forceps
Merit™ Sterile Kelly Hemostatic Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue.
  • Ready-to-use
  • Available straight or curved
  • Ratcheted finger ring handle
The forceps may also be used for soft tissue dissection. They are available curved or straight with a ratcheted finger ring handle. The jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws. Merit™ Sterile instruments are ready-to-use, sterile, disposable, office-grade instruments sold in bulk quantities.