Foerster Sponge Forceps, OR Grade, Sklar
Supplier: Sklar
87-2195 87-2395 87-2095 87-2295 87-2490
10193-660EA 191.36 CAD
10193-660 10193-764 10199-040 10199-044 10015-652
Foerster Sponge Forceps, OR Grade, Sklar
Forceps
Censitrac Ready products are industry standard and constructed of stainless steel.
  • Foerster Sponge Forceps, also known as Foerster-Ballenger Sponge Forceps, are commonly used in surgical procedures to hold gauze squares that will absorb excess fluids and blood from the surgical site or hemostasis. The ratcheted locking mechanism of these finger ring forceps securely grip the gauze and hold it in place. Foerster sponge forceps are often used in bariatric surgeries because of their longer length, and the curved version may be used for removing the placenta in OB/GYN procedures.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.