Mayo Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
15-1555 15-1567 15-2555 15-2567 90-1690 90-1590
82029-944EA 166.66 CAD
82029-944 82029-946 82029-958 82029-960 10192-314 10192-384
Mayo Dissecting Scissors, OR Grade, Sklar
Scissors
Mayo Dissecting Scissors are a versatile instrument, that can be used in multiple procedures and settings.
They are available straight or curved, in a variety of lengths, and have a standard beveled blade
Censitrac Ready products are industry standard and constructed of stainless steel.
While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference. Sklar also offers Mayo DissectingMore Product Information