Metzenbaum-Nelson Dissecting Scissors, OR Grade, Sklar®
Supplier: Sklar
55-9080 55-9084 55-9180 55-9184
10193-906EA 344.81 CAD
10193-906 10197-670 10197-674 10197-676
Metzenbaum-Nelson Dissecting Scissors, OR Grade, Sklar®
Scissors
Metzenbaum-Nelson Dissecting Scissors are used for dissecting and cutting tissue.
These scissors are not recommended for cutting sutures, drains, or heavy tissue
Censitrac Ready products are industry standard and constructed of stainless steel.
Metzenbaum-Nelson scissors tend to be longer than standard Metzenbaum scissors and are ideal for use in thoracic procedures.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.