Metzenbaum-Nelson Bariatric Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
55-9090 55-9311 55-9094 55-9194 55-9210 55-9310 55-9411
10192-388EA 350.21 CAD
10192-388 10194-436 10197-672 10197-678 10197-680 10197-682 10197-684
Metzenbaum-Nelson Bariatric Dissecting Scissors, OR Grade, Sklar
Scissors
Metzenbaum-Nelson Dissecting Scissors are used for dissecting and cutting tissue.
These scissors are not recommended for cutting sutures, drains, or heavy tissue
Censitrac Ready products are industry standard and constructed of stainless steel.
Metzenbaum-Nelson scissors tend to be longer than standard Metzenbaum scissors and are ideal for use in thoracic procedures.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
More Product Information