Doyen Scissors, OR Grade, Sklar
Supplier: Sklar
15-1307 15-1308 15-2307 15-2308
89025-492EA 318.47 CAD
89025-492 89025-494 89025-506 10195-006
Doyen Scissors, OR Grade, Sklar
Scissors
Sklar Doyen Scissors, which may also be known as Doyen Abdominal Scissors, are versatile, multipurpose scissors used for both dissection and cutting in many surgical procedures
They are heavier than standard Mayo scissors.
Order Now
SPECIFICATIONS
Environmentally Preferable