Sklarlite™ Operating Scissors, OR Grade, Sklar
Supplier: Sklar
23-1100 23-1101 23-1102 23-1104 23-1136 23-1138 23-1140 23-1105 23-1106 23-1113 23-1124 23-1126 23-1128 23-1130
10195-068EA 303.69 CAD
10195-068 10195-070 10195-072 10195-074 10195-076 10195-078 10195-080 10195-252 10195-254 10195-258 10195-268 10195-270 10195-272 10195-274
Sklarlite™ Operating Scissors, OR Grade, Sklar
Scissors
Sklarlite™ Operating Scissors, also known as Surgical Scissors are a versatile surgical instrument.
With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying Blade Types and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze.
Sklarlite™ instruments are a lighter version of standard Sklar instruments. A unique manufacturing process developed by Sklar shaves microns off standard and delicate pattern instruments while maintaining structural integrity and durability. This lightweight modification provides the surgeon with greater control and less fatigue.