Double Edge Razor Blade, Pathco
Supplier: CANCER DIAGNOSTICS, INC.
D2877 D28782C
25861-010EA 484.21 CAD
25861-010 25861-012
Double Edge Razor Blade, Pathco
Razors and Blades
The disposable, double-edge razor blade is 5.08 cm (2") long, straight, rigid and can be locked into position with a few twists of the knurled knob.
The handle is made of hard-anodized aluminum and designed by pathologists to aid in cutting tissues. It can be autoclaved.