Leakproof HDPE Bottles, Natural, C&G Containers
Supplier: C&G Containers
LPV008900000N LPV008900102 LPV016200062 LPV032200055
71001-064CS 1323.75 CAD
71001-064 71001-068 71001-074 71001-090 71001-096 89125-422
Leakproof HDPE Bottles, Natural, C&G Containers
Bottles Water Sampling Bottles
These plastic bottles are used for the collection of aqueous samples and for testing metals and a variety of inorganic parameters, including alkalinity, anions, acidity, cyanide, sulfate, and hardness.
Pre-cleaned and Certified products contain the Certificate of Analysis and cleaning protocol form in the product case.
A lot certificate of analysis is generated with the analytical results obtained including the concentration reporting limits and units of measurement, for Pre-cleaned and Certified products. This certificate of analysis is included in each product case.
Wash protocols for these bottles include; A1 (Nitric acid wash), B1 (Deionized water wash), C1 (Hydrochloric acid wash), and L3 (no wash). Some bottles are chemically preserved.
Not all analytical parameters are included on the Certificate of Analysis. Please refer to the Certificate of Analysis for certified analytes and corresponding reporting limits. Secondary uses for these containers include containment and archiving.
Ordering information: Container measurements and capacity "contain" volumes may vary.