Dropette® Disposable Transfer Pipettes, Simport
Supplier: Simport Scientific
P200-5610S P200-521S P200-5220S P200-14 P200-60 P200-82 P200-58 P200-44 P200-10 P200-14 P200-20 P200-30 P200-44 P200-52 P200-56 P200-58 P200-58V P200-60 P200-72 P200-82 P200-521S P200-5220S P200-5610S
CA60872-447CS 1395.6 CAD
CA60872-447 CA60830-000 CA60872-450 CA60872-456 CA60872-458 CA11014-044 CA11014-048 CA11014-050 CA11016-300 CA11016-302 10118-348 10118-350 10118-352 10118-354 10118-356 10118-358 10118-360 10118-362 10118-364 10118-366 10118-368 10118-370 76299-266 76299-268 76299-270 76299-272 76299-274
Dropette® Disposable Transfer Pipettes, Simport
Pipettes Pasteur/Transfer Pipettes
These unbreakable all-in-one pipettes eliminate the hazard of broken glass and exposure to infectious materials.
  • Can be sealed and refrigerated
  • Will not shatter or break
  • Non toxic and inert and could be used in liquid nitrogen
  • Uniform drop size having safe transfer of fluids
  • Temperature resistant down to –196 °C
  • Seven models provide graduations
Rubber bulbs are attached with glass pipettes and molded from see-through low density polyethylene. The low-affinity surface reduces the loss of cells and valuable proteins due to binding. Inert to biologicalMore Product Information