Probes with Eye, OR Grade, Sklar
Supplier: Sklar
30-1945 30-1980 30-2010
10195-462EA 53.66 CAD
10195-462 10195-460 10195-458
Probes with Eye, OR Grade, Sklar
Dissection Needles
These Probes with Eye may be used to explore or gauge the depth of non-visible cavities, explore lesions, or manipulate tissue
They are a slender, slightly malleable, multipurpose instrument with a bulbous tip that can be used in a variety of procedures.