Probe with Eye, OR Grade, Sklar
Supplier: Sklar
30-1950 30-1960 30-2012 30-1955
10192-262EA 40.17 CAD
10192-262 10192-416 10193-330 10193-740
Probe with Eye, OR Grade, Sklar
Dissection Needles
Probe with Eye is a slender, malleable, multipurpose instrument with a bulbous tip that can be used in a variety of procedures.
It may be used to explore or gauge the depth of non-visible cavities, explore lesions, or manipulate tissue. The eye may be used for carrying ligatures.